siRNA Details
| Gene Symbol: Gpatch2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027405.2 | NP_081681.2 | 9 | 1334 | ||
| GenBank mRNA | AK129299.1 | - | 9 | 1311 | ||
| AK181673.1 | - | 1 | 45 | |||
| AK203506.1 | - | 1 | 45 | |||
| BC048169.1 | - | 9 | 1343 | 6821902 | ||
| BC050782.1 | - | 9 | 1314 | |||
| BC058256.1 | - | 9 | 1334 | 6466603 | ||

Gpatch2l
Genomic Map