siRNA Details
| Gene Symbol: Derl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024440.2 | NP_077760.1 | 5 | 455 | ||
| GenBank mRNA | AB047555.1 | - | 5 | 455 | ||
| AK007948.1 | - | 5 | 441 | |||
| AK172429.1 | - | 5 | 441 | |||
| BC004729.1 | - | 5 | 438 | |||
| BC019815.1 | - | 5 | 435 | |||
| BC099615.1 | - | 5 | 435 | |||
| BC115466.1 | - | 5 | 450 | 40100387 | ||
| BC115467.1 | - | 5 | 450 | 40100388 | ||

Derl3
Genomic Map