siRNA Details
| Gene Symbol: Derl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024440.2 | NP_077760.1 | 4 | 313 | ||
| GenBank mRNA | AB047555.1 | - | 4 | 313 | ||
| AK007348.1 | - | 4 | 298 | |||
| AK007948.1 | - | 4 | 299 | |||
| AK172429.1 | - | 4 | 299 | |||
| BC004729.1 | - | 4 | 296 | |||
| BC019815.1 | - | 4 | 292 | |||
| BC099615.1 | - | 4 | 293 | |||
| BC115466.1 | - | 4 | 308 | 40100387 | ||
| BC115467.1 | - | 4 | 308 | 40100388 | ||

Derl3
Genomic Map