siRNA Details
| Gene Symbol: Cox10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178379.3 | NP_848466.1 | 5 | 736 | ||
| GenBank mRNA | AK010385.1 | - | 5 | 730 | ||
| AK149999.1 | - | 5 | 726 | |||
| AK154668.1 | - | 5 | 735 | |||
| AK171661.1 | - | 5 | 726 | |||
| AK178210.1 | - | 2 | 58 | |||
| AK184676.1 | - | 2 | 160 | |||
| BC018301.1 | - | 5 | 687 | |||
| BC038046.1 | - | 5 | 689 | 4206826 | ||

Cox10
Genomic Map