siRNA Details
| Gene Symbol: Cox10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178379.3 | NP_848466.1 | 3 | 533 | ||
| GenBank mRNA | AK010385.1 | - | 3 | 527 | ||
| AK149999.1 | - | 3 | 523 | |||
| AK154668.1 | - | 3 | 532 | |||
| AK156865.1 | - | 3 | 532 | |||
| AK171661.1 | - | 3 | 523 | |||
| AK172718.1 | - | 3 | 532 | |||
| BC018301.1 | - | 3 | 484 | |||
| BC038046.1 | - | 3 | 486 | 4206826 | ||

Cox10
Genomic Map