siRNA Details
| Gene Symbol: Asb12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080858.3 | NP_543134.1 | 2 | 889 | ||
| GenBank mRNA | AF403040.1 | - | 2 | 890 | ||
| AK009644.1 | - | 2 | 890 | |||
| BC107257.2 | - | 2 | 854 | 40052462 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asb12| Gene Symbol: Asb12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080858.3 | NP_543134.1 | 2 | 889 | ||
| GenBank mRNA | AF403040.1 | - | 2 | 890 | ||
| AK009644.1 | - | 2 | 890 | |||
| BC107257.2 | - | 2 | 854 | 40052462 | ||