siRNA Details
| Gene Symbol: Kptn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133727.2 | NP_598488.2 | 3 | 417 | ||
| GenBank mRNA | AK009746.1 | - | 3 | 398 | ||
| AK081226.1 | - | 2 | 548 | |||
| BC018316.1 | - | 3 | 385 | 4160465 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kptn| Gene Symbol: Kptn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133727.2 | NP_598488.2 | 3 | 417 | ||
| GenBank mRNA | AK009746.1 | - | 3 | 398 | ||
| AK081226.1 | - | 2 | 548 | |||
| BC018316.1 | - | 3 | 385 | 4160465 | ||