siRNA Details
| Gene Symbol: Stk26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133729.1 | NP_598490.1 | 3 | 462 | ||
| GenBank mRNA | AK034609.1 | - | 3 | 501 | ||
| BC005708.1 | - | 3 | 462 | 2598933 | ||
| BC132438.1 | - | 2 | 265 | 40130715 | ||
| BC138193.1 | - | 2 | 265 | 8861212 | ||
| BC145189.1 | - | 2 | 265 | |||

Stk26
Genomic Map