siRNA Details
Gene Symbol: Csnk1g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152809.2 | NP_690022.2 | 5 | 1100 | ||
GenBank mRNA | AK218008.1 | - | 2 | 150 | ||
AK218677.1 | - | 2 | 126 | |||
BC033601.1 | - | 2 | 115 | |||
BQ443928.1 | - | 2 | 134 | |||
CN454971.1 | - | 4 | 583 |
