siRNA Details
| Gene Symbol: Mier2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027422.2 | NP_081698.2 | 11, 12 | 1114 | ||
| GenBank mRNA | AK154672.1 | - | 11, 12 | 1131 | ||
| AK173128.1 | - | 10, 11 | 1270 | |||
| BC067013.1 | - | 11, 12 | 1114 | 6810181 | ||
| BC080757.1 | - | 4, 5 | 365 | |||
| BC083321.1 | - | 2, 3 | 235 | |||

Mier2
Genomic Map