siRNA Details
| Gene Symbol: Mier2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027422.2 | NP_081698.2 | 9 | 868 | ||
| GenBank mRNA | AK154672.1 | - | 9 | 885 | ||
| AK173128.1 | - | 8 | 1024 | |||
| BC067013.1 | - | 9 | 868 | 6810181 | ||
| BC080757.1 | - | 2 | 119 | |||

Mier2
Genomic Map