siRNA Details
| Gene Symbol: Polr3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027423.1 | NP_081699.1 | 27 | 3193 | ||
| GenBank mRNA | AK012532.1 | - | 2 | 102 | ||
| BC044796.1 | - | 4 | 381 | |||
| BC068143.1 | - | 27 | 3193 | 30362434 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Polr3b| Gene Symbol: Polr3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027423.1 | NP_081699.1 | 27 | 3193 | ||
| GenBank mRNA | AK012532.1 | - | 2 | 102 | ||
| BC044796.1 | - | 4 | 381 | |||
| BC068143.1 | - | 27 | 3193 | 30362434 | ||