siRNA Details
| Gene Symbol: Draxin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027426.3 | NP_081702.2 | 4 | 862 | ||
| GenBank mRNA | AK011831.1 | - | 1 | 27 | ||
| AK086198.1 | - | 4 | 863 | |||
| BC060239.1 | - | 4 | 843 | 6853328 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Draxin| Gene Symbol: Draxin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027426.3 | NP_081702.2 | 4 | 862 | ||
| GenBank mRNA | AK011831.1 | - | 1 | 27 | ||
| AK086198.1 | - | 4 | 863 | |||
| BC060239.1 | - | 4 | 843 | 6853328 | ||