siRNA Details
| Gene Symbol: Draxin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027426.3 | NP_081702.2 | 6 | 1021 | ||
| GenBank mRNA | AB301918.1 | - | 5 | 886 | ||
| AK011831.1 | - | 3 | 186 | |||
| AK086198.1 | - | 6 | 1022 | |||
| BC058734.1 | - | 2 | 137 | |||
| BC060239.1 | - | 6 | 1002 | 6853328 | ||

Draxin
Genomic Map