siRNA Details
| Gene Symbol: Inf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198411.2 | NP_940803.2 | 3 | 530 | ||
| GenBank mRNA | CD347830.1 | - | 2 | 163 | ||
| DQ834374.1 | - | 3 | 532 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Inf2| Gene Symbol: Inf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198411.2 | NP_940803.2 | 3 | 530 | ||
| GenBank mRNA | CD347830.1 | - | 2 | 163 | ||
| DQ834374.1 | - | 3 | 532 | |||