siRNA Details
| Gene Symbol: Dhrs13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183286.2 | NP_899109.2 | 5 | 1141 | ||
| GenBank mRNA | AK011939.1 | - | 5 | 1112 | ||
| AK046465.1 | - | 4 | 1188 | |||
| BC115881.1 | - | 4 | 986 | 40088339 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhrs13| Gene Symbol: Dhrs13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183286.2 | NP_899109.2 | 5 | 1141 | ||
| GenBank mRNA | AK011939.1 | - | 5 | 1112 | ||
| AK046465.1 | - | 4 | 1188 | |||
| BC115881.1 | - | 4 | 986 | 40088339 | ||