siRNA Details
| Gene Symbol: 2610318N02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183287.2 | NP_899110.2 | 5, 6 | 872 | ||
| GenBank mRNA | AK012048.1 | - | 4, 5 | 753 | ||
| AK146836.1 | - | 4, 5 | 764 | |||
| BC039993.1 | - | 5, 6 | 872 | 1446463 | ||

2610318N02Rik
Genomic Map