siRNA Details
| Gene Symbol: Crtc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173863.2 | NP_776288.2 | 14 | 1736 | ||
| GenBank mRNA | AK144840.1 | - | 14 | 1739 | ||
| BC011210.1 | - | 14 | 1575 | 4195538 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crtc3| Gene Symbol: Crtc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173863.2 | NP_776288.2 | 14 | 1736 | ||
| GenBank mRNA | AK144840.1 | - | 14 | 1739 | ||
| BC011210.1 | - | 14 | 1575 | 4195538 | ||