siRNA Details
| Gene Symbol: Atad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027435.2 | NP_081711.2 | 18 | 2396 | ||
| GenBank mRNA | AK029867.1 | - | 11 | 1543 | ||
| AK134319.1 | - | 5 | 772 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atad2| Gene Symbol: Atad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027435.2 | NP_081711.2 | 18 | 2396 | ||
| GenBank mRNA | AK029867.1 | - | 11 | 1543 | ||
| AK134319.1 | - | 5 | 772 | |||