siRNA Details
| Gene Symbol: Atp6ap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027439.4 | NP_081715.1 | 2 | 225 | ||
| GenBank mRNA | AB192471.2 | - | 2 | 101 | ||
| AK017482.1 | - | 2 | 157 | |||
| AK029405.1 | - | 2 | 128 | |||
| AK076495.1 | - | 2 | 170 | |||
| AK151735.1 | - | 2 | 156 | |||
| AK168139.1 | - | 2 | 184 | |||
| AK210696.1 | - | 1 | 44 | |||
| BC014706.1 | - | 2 | 96 | 4457079 | ||
| BY784908.1 | - | 2 | 226 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001007091, AB188298, BC079339.

Atp6ap2
Genomic Map