siRNA Details
| Gene Symbol: Tmem242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027457.4 | NP_081733.3 | 4 | 400 | ||
| GenBank mRNA | AK009851.1 | - | 4 | 383 | ||
| AK043276.1 | - | 4 | 368 | |||
| AK088025.1 | - | 4 | 380 | |||
| AK155326.1 | - | 4 | 379 | |||
| AK158107.1 | - | 4 | 379 | |||
| BC019420.1 | - | 4 | 364 | 5053733 | ||

Tmem242
Genomic Map