siRNA Details
| Gene Symbol: Tln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081242.2 | NP_001074711.2 | 50 | 6789 | ||
| GenBank mRNA | AB093231.1 | - | 24 | 3331 | ||
| AF467081.1 | - | 46 | 6248 | |||
| AK017597.1 | - | 4 | 561 | |||
| AK029828.1 | - | 25 | 3482 | |||
| AK040595.1 | - | 4 | 559 | |||
| BC033288.1 | - | 12 | 1685 | |||
| BC059856.1 | - | 4 | 550 | |||
| BC079896.1 | - | 35 | 4652 | 30544295 | ||
| BC158057.1 | - | 50 | 6760 | 9007357 | ||

Tln2
Genomic Map