siRNA Details
| Gene Symbol: Tmtc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028651.2 | NP_082927.1 | 6 | 769 | ||
| GenBank mRNA | AK028333.1 | - | 6 | 750 | ||
| AK033947.1 | - | 6 | 770 | |||
| AK036741.1 | - | 6 | 726 | |||
| AK039093.1 | - | 6 | 714 | |||
| AK043293.1 | - | 6 | 796 | |||
| AK045271.1 | - | 6 | 731 | |||
| AK049748.1 | - | 7 | 910 | |||
| AK082526.1 | - | 5 | 597 | |||
| BC031368.1 | - | 2 | 143 | 5029461 | ||

Tmtc4
Genomic Map