siRNA Details
| Gene Symbol: Tmtc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028651.2 | NP_082927.1 | 10 | 1348 | ||
| GenBank mRNA | AK028333.1 | - | 10 | 1329 | ||
| AK033947.1 | - | 10 | 1349 | |||
| AK036741.1 | - | 9 | 1124 | |||
| AK039093.1 | - | 10 | 1294 | |||
| AK043293.1 | - | 10 | 1375 | |||
| AK044391.1 | - | 10 | 1304 | |||
| AK045271.1 | - | 10 | 1310 | |||
| AK049748.1 | - | 11 | 1489 | |||
| AK082526.1 | - | 9 | 1176 | |||
| BC031368.1 | - | 6 | 722 | 5029461 | ||

Tmtc4
Genomic Map