siRNA Details
| Gene Symbol: Tmtc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028651.2 | NP_082927.1 | 2 | 291 | ||
| GenBank mRNA | AK028333.1 | - | 2 | 272 | ||
| AK033947.1 | - | 2 | 292 | |||
| AK036741.1 | - | 2 | 248 | |||
| AK039093.1 | - | 2 | 236 | |||
| AK043293.1 | - | 2 | 318 | |||
| AK044391.1 | - | 2 | 247 | |||
| AK045271.1 | - | 2 | 253 | |||
| AK049748.1 | - | 3 | 432 | |||
| AK082526.1 | - | 1 | 119 | |||

Tmtc4
Genomic Map