siRNA Details
| Gene Symbol: Arhgap27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183288.3 | NP_899111.2 | 3 | 1344 | ||
| GenBank mRNA | AK017636.1 | - | 3 | 1034 | ||
| BC138292.1 | - | 1 | 800 | 8861311 | ||
| BC138293.1 | - | 1 | 799 | 8861312 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgap27| Gene Symbol: Arhgap27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183288.3 | NP_899111.2 | 3 | 1344 | ||
| GenBank mRNA | AK017636.1 | - | 3 | 1034 | ||
| BC138292.1 | - | 1 | 800 | 8861311 | ||
| BC138293.1 | - | 1 | 799 | 8861312 | ||