siRNA Details
| Gene Symbol: Pak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027470.3 | NP_081746.1 | 5 | 1267 | ||
| GenBank mRNA | AK088512.1 | - | 5 | 1244 | ||
| AK129298.2 | - | 5 | 1263 | |||
| AY217016.1 | - | 3 | 982 | |||
| BC030389.1 | - | 2 | 425 | |||
| BC048238.1 | - | 5 | 1229 | 6475114 | ||
| BQ572351.1 | - | 2 | 525 | |||

Pak4
Genomic Map