siRNA Details
| Gene Symbol: Ssfa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080558.4 | NP_542125.3 | 8 | 986 | ||
| GenBank mRNA | AB093303.1 | - | 8 | 861 | ||
| AB120565.1 | - | 8 | 871 | |||
| AK037213.1 | - | 6 | 696 | |||
| AK169543.1 | - | 8 | 987 | |||
| BC079535.1 | - | 2 | 365 | |||
| BC122519.1 | - | 8 | 872 | |||
| BC141883.1 | - | 8 | 871 | 40097831 | ||

Ssfa2
Genomic Map