siRNA Details
| Gene Symbol: Ankrd24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027480.3 | NP_081756.4 | 4, 5 | 519 | ||
| GenBank mRNA | AK043638.1 | - | 4, 5 | 353 | ||
| AK173316.1 | - | 4, 5 | 342 | |||
| BC048236.1 | - | 3, 4 | 424 | 6313999 | ||
| CB524981.1 | - | 1, 2 | 48 | |||
| CJ177454.1 | - | 4, 5 | 358 | |||

Ankrd24
Genomic Map