siRNA Details
| Gene Symbol: Ankrd24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027480.3 | NP_081756.4 | 17 | 1782 | ||
| GenBank mRNA | AK019475.1 | - | 4 | 448 | ||
| AK043638.1 | - | 15 | 2864 | |||
| AK173316.1 | - | 15 | 2748 | |||
| BC017633.1 | - | 4 | 453 | |||
| BC048236.1 | - | 16 | 1687 | 6313999 | ||

Ankrd24
Genomic Map