siRNA Details
| Gene Symbol: Oip5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042653.1 | NP_001036118.1 | 5 | 652 | ||
| GenBank mRNA | AK017825.1 | - | 5 | 617 | ||
| AK045323.1 | - | 4 | 855 | |||
| BC115883.1 | - | 5 | 608 | 40088361 | ||
| BC116660.1 | - | 5 | 609 | 40088363 | ||

Oip5
Genomic Map