siRNA Details
| Gene Symbol: Zcchc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027494.3 | NP_081770.3 | 3 | 409 | ||
| GenBank mRNA | AK017857.1 | - | 3 | 305 | ||
| AK032349.1 | - | 3 | 387 | |||
| AK033469.1 | - | 3 | 359 | |||
| AK156770.1 | - | 3 | 381 | |||
| AK166990.1 | - | 3 | 316 | |||
| AK167126.1 | - | 3 | 314 | |||
| AK167736.1 | - | 3 | 316 | |||
| BC013555.1 | - | 2 | 61 | 4238634 | ||

Zcchc8
Genomic Map