siRNA Details
| Gene Symbol: Fam175a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172405.3 | NP_765993.1 | 5 | 433 | ||
| GenBank mRNA | AK014420.1 | - | 5 | 459 | ||
| AK042001.1 | - | 5 | 442 | |||
| AK042339.1 | - | 5 | 442 | |||
| AK051816.1 | - | 5 | 434 | |||
| AK167540.1 | - | 5 | 464 | |||
| BC029845.1 | - | 5 | 438 | 4010744 | ||
| BC043923.1 | - | 4 | 347 | |||

Fam175a
Genomic Map