siRNA Details
| Gene Symbol: 3830403N18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027510.2 | NP_081786.1 | 5, 6 | 401 | ||
| GenBank mRNA | AK014416.1 | - | 5, 6 | 402 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3830403N18Rik| Gene Symbol: 3830403N18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027510.2 | NP_081786.1 | 5, 6 | 401 | ||
| GenBank mRNA | AK014416.1 | - | 5, 6 | 402 | ||