siRNA Details
| Gene Symbol: Hmha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142701.1 | NP_001136173.1 | 3, 4 | 724 | ||
| NM_027521.3 | NP_081797.1 | 2, 3 | 276 | |||
| GenBank mRNA | AK044860.1 | - | 2, 3 | 238 | ||
| AK151423.1 | - | 3, 4 | 668 | |||
| AK159194.1 | - | 3, 4 | 725 | |||
| AK159474.1 | - | 3, 4 | 657 | |||
| AK170262.1 | - | 3, 4 | 710 | |||
| AK171304.1 | - | 3, 4 | 710 | |||
| BC053750.1 | - | 2, 3 | 276 | 6333971 | ||

Hmha1
Genomic Map