siRNA Details
| Gene Symbol: Ggnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001251881.1 | NP_001238810.1 | 5 | 662 | ||
| NM_027544.2 | NP_081820.2 | 4 | 1091 | |||
| GenBank mRNA | AB250379.1 | - | 4 | 974 | ||
| AK077129.1 | - | 4 | 974 | |||
| AY141192.1 | - | 4 | 1091 | |||
| BC049764.1 | - | 4 | 952 | 6774330 | ||
| CF585840.1 | - | 5 | 674 | |||

Ggnbp1
Genomic Map