siRNA Details
| Gene Symbol: Prdm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027547.2 | NP_081823.2 | 3 | 360 | ||
| GenBank mRNA | AK014501.1 | - | 3 | 284 | ||
| AK018310.1 | - | 3 | 298 | |||
| AK021384.1 | - | 2 | 219 | |||
| AK183711.1 | - | 1 | 41 | |||
| BC089598.1 | - | 3 | 300 | |||
| BC138901.1 | - | 3 | 334 | 8861923 | ||
| BC138902.1 | - | 3 | 334 | 8861924 | ||
| BY014014.1 | - | 3 | 361 | |||

Prdm5
Genomic Map