siRNA Details
| Gene Symbol: Prdm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027547.2 | NP_081823.2 | 14, 15 | 1767 | ||
| GenBank mRNA | AK014501.1 | - | 14, 15 | 1691 | ||
| AK018310.1 | - | 10, 11 | 1153 | |||
| AK021384.1 | - | 5, 6 | 576 | |||
| BC138901.1 | - | 14, 15 | 1741 | 8861923 | ||
| BC138902.1 | - | 14, 15 | 1741 | 8861924 | ||
| BX511446.1 | - | 3, 4 | 270 | |||

Prdm5
Genomic Map