siRNA Details
| Gene Symbol: Klhl30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027551.2 | NP_081827.1 | 7 | 1562 | ||
| GenBank mRNA | AK014532.1 | - | 3 | 429 | ||
| AK086025.1 | - | 7 | 1563 | |||
| AK146580.1 | - | 7 | 1619 | |||
| AK148398.1 | - | 7 | 1571 | |||
| BC115428.1 | - | 6 | 1505 | 40094085 | ||
| BC115429.1 | - | 6 | 1505 | 40094088 | ||

Klhl30
Genomic Map