siRNA Details
| Gene Symbol: Ankib1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003909.4 | NP_001003909.2 | 5 | 1315 | ||
| NM_001289527.1 | NP_001276456.1 | 5 | 1315 | |||
| NM_001289529.1 | NP_001276458.1 | 4 | 787 | |||
| GenBank mRNA | AK014523.1 | - | 6 | 1077 | ||
| AK129343.1 | - | 5 | 1116 | |||
| BC020475.1 | - | 5 | 849 | |||
| BC089031.1 | - | 4 | 766 | |||
| BC145511.1 | - | 5 | 1066 | 9054074 | ||
| BC150792.1 | - | 5 | 1066 | 9087703 | ||

Ankib1
Genomic Map