siRNA Details
| Gene Symbol: Cep192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027556.1 | NP_081832.1 | 22 | 4549 | ||
| GenBank mRNA | AK122526.1 | - | 3 | 1112 | ||
| BC064462.1 | - | 3 | 595 | |||
| CK639048.1 | - | 1 | 38 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep192| Gene Symbol: Cep192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027556.1 | NP_081832.1 | 22 | 4549 | ||
| GenBank mRNA | AK122526.1 | - | 3 | 1112 | ||
| BC064462.1 | - | 3 | 595 | |||
| CK639048.1 | - | 1 | 38 | |||