siRNA Details
| Gene Symbol: Pgrmc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027558.1 | NP_081834.1 | 2 | 528 | ||
| GenBank mRNA | AK014543.1 | - | 2 | 427 | ||
| AK140064.1 | - | 2 | 508 | |||
| AK184727.1 | - | 1 | 78 | |||
| BC044759.2 | - | 2 | 469 | |||
| CD363842.1 | - | 2 | 527 | |||

Pgrmc2
Genomic Map