siRNA Details
| Gene Symbol: Cfap43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027559.2 | NP_081835.2 | 36 | 4736 | ||
| GenBank mRNA | BC107402.1 | - | 13 | 1478 | ||
| BC107403.1 | - | 16 | 1584 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfap43| Gene Symbol: Cfap43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027559.2 | NP_081835.2 | 36 | 4736 | ||
| GenBank mRNA | BC107402.1 | - | 13 | 1478 | ||
| BC107403.1 | - | 16 | 1584 | |||