siRNA Details
Gene Symbol: Cfap43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027559.2 | NP_081835.2 | 34 | 4474 | ||
GenBank mRNA | BC107402.1 | - | 11 | 1216 | ||
BC107403.1 | - | 11 | 1216 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cfap43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027559.2 | NP_081835.2 | 34 | 4474 | ||
GenBank mRNA | BC107402.1 | - | 11 | 1216 | ||
BC107403.1 | - | 11 | 1216 |