siRNA Details
| Gene Symbol: Krtap31-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027568.2 | NP_081844.1 | 1 | 913 | ||
| GenBank mRNA | AK014635.1 | - | 1 | 914 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap31-1| Gene Symbol: Krtap31-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027568.2 | NP_081844.1 | 1 | 913 | ||
| GenBank mRNA | AK014635.1 | - | 1 | 914 | ||