siRNA Details
| Gene Symbol: Prss22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133731.2 | NP_598492.2 | 3 | 366 | ||
| GenBank mRNA | AB010778.1 | - | 3 | 349 | ||
| AK014645.1 | - | 3 | 366 | |||
| BC055854.1 | - | 3 | 351 | 4035351 | ||
| CD773447.1 | - | 3 | 370 | |||

Prss22
Genomic Map