siRNA Details
| Gene Symbol: P2ry12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027571.3 | NP_081847.3 | 4 | 1028 | ||
| GenBank mRNA | AJ312130.1 | - | 1 | 675 | ||
| AK013804.1 | - | 4 | 1029 | |||
| AK014807.1 | - | 3 | 882 | |||
| BC025428.1 | - | 5 | 1093 | |||
| BC027381.1 | - | 3 | 888 | 4947057 | ||
More Information
Set Membership: |
|

P2ry12
Genomic Map