siRNA Details
| Gene Symbol: Armc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081083.2 | NP_001074552.1 | 13 | 1927 | ||
| NM_001271563.1 | NP_001258492.1 | 13 | 1927 | |||
| NM_001271564.1 | NP_001258493.1 | 13 | 1927 | |||
| NM_001271565.1 | NP_001258494.1 | 13 | 1927 | |||
| GenBank mRNA | AK014880.1 | - | 13 | 1743 | ||
| AK041744.1 | - | 13 | 1751 | |||
| BC145004.1 | - | 13 | 1735 | |||
| BC145005.1 | - | 13 | 1735 | 9053553 | ||
| BC145013.1 | - | 13 | 1735 | 9053561 | ||
| BC150688.1 | - | 13 | 1735 | 9087599 | ||

Armc3
Genomic Map