siRNA Details
| Gene Symbol: Dmrtc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038616.2 | NP_001033705.2 | 2, 3 | 338 | ||
| NM_027591.3 | NP_081867.1 | 4, 5 | 436 | |||
| NM_029378.2 | NP_083654.1 | 2, 3 | 250 | |||
| GenBank mRNA | AK006548.1 | - | 2, 3 | 353 | ||
| AK014934.1 | - | 4, 5 | 437 | |||
| BC147175.1 | - | 4, 5 | 408 | 9056223 | ||
| BC147176.1 | - | 4, 5 | 408 | 9056216 | ||
| DQ316772.1 | - | 4, 5 | 392 | |||
| DQ316773.1 | - | 2, 3 | 250 | |||
| DQ316774.1 | - | 2, 3 | 250 | |||

Dmrtc1a
Genomic Map