siRNA Details
| Gene Symbol: Ttll7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014905.1 | - | 12 | 1696 | ||
| AM690750.1 | - | 11 | 1337 | |||
| AM690751.1 | - | 11 | 1337 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttll7| Gene Symbol: Ttll7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014905.1 | - | 12 | 1696 | ||
| AM690750.1 | - | 11 | 1337 | |||
| AM690751.1 | - | 11 | 1337 | |||